Assay Details
| Gene Symbol: Cdhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130878.2 | NP_570948.1 | 3 - 4 | 471 | 80 | |
| GenBank mRNA | AB093300.1 | - | 3 - 4 | 887 | 80 | |
| AF426393.1 | - | 3 - 4 | 358 | 80 | ||
| AK143151.1 | - | 3 - 4 | 410 | 80 | ||
| BC042454.1 | - | 3 - 4 | 381 | 5369576 | 80 | |
| BC058270.1 | - | 3 - 4 | 471 | 6490597 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdhr1
Genomic Map