Assay Details
| Gene Symbol: Sh3tc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194344.2 | NP_919325.2 | 9 - 10 | 1507 | 89 | |
| GenBank mRNA | AK028482.1 | - | 5 - 6 | 1603 | 89 | |
| AK150064.1 | - | 9 - 10 | 1285 | 89 | ||
| AK155330.1 | - | 9 - 10 | 1268 | 90 | ||
| AK159089.1 | - | 9 - 10 | 1286 | 89 | ||
| AK167283.1 | - | 9 - 10 | 1208 | 89 | ||
| AK170383.1 | - | 9 - 10 | 1340 | 89 | ||
| AK170396.1 | - | 9 - 10 | 1287 | 89 | ||
| AK171209.1 | - | 9 - 10 | 1264 | 89 | ||
| AK171242.1 | - | 9 - 10 | 1264 | 89 | ||
| BC024909.1 | - | 7 - 8 | 957 | 89 | ||
| BC056461.1 | - | 7 - 8 | 851 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3tc1
Genomic Map