Assay Details
| Gene Symbol: Sptssb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164210.2 | NP_001157682.1 | 2 - 3 | 101 | 96 | |
| GenBank mRNA | AK009973.1 | - | 2 - 3 | 68 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK009973.1, NM_001164210.2 transcripts.
Set Membership: |
|

Sptssb
Genomic Map