Assay Details
| Gene Symbol: Stim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081103.2 | NP_001074572.2 | 10 - 11 | 1853 | 83 | |
| GenBank mRNA | AK041804.1 | - | 4 - 5 | 407 | 83 | |
| AK173183.1 | - | 10 - 11 | 1853 | 83 | ||
| AM712359.1 | - | 10 - 11 | 1492 | 83 | ||
| BC043455.1 | - | 3 - 4 | 375 | 83 | ||
| BC137881.1 | - | 10 - 11 | 1759 | 8860900 | 83 | |
| BC145001.1 | - | 10 - 11 | 1761 | 9053549 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stim2
Genomic Map