Assay Details
| Gene Symbol: Stim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081103.2 | NP_001074572.2 | 5 - 6 | 990 | 98 | |
| GenBank mRNA | AK080310.1 | - | 5 - 6 | 948 | 98 | |
| AK173183.1 | - | 5 - 6 | 990 | 98 | ||
| AM712359.1 | - | 5 - 6 | 629 | 98 | ||
| BC137881.1 | - | 5 - 6 | 896 | 8860900 | 98 | |
| BC145001.1 | - | 5 - 6 | 898 | 9053549 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stim2
Genomic Map