Assay Details
| Gene Symbol: Ufl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026194.4 | NP_080470.2 | 13 - 14 | 1579 | 102 | |
| GenBank mRNA | AK007993.2 | - | 13 - 14 | 1524 | 102 | |
| AK013382.1 | - | 6 - 7 | 841 | 102 | ||
| AK032663.1 | - | 13 - 14 | 1515 | 102 | ||
| AK076206.1 | - | 13 - 14 | 1520 | 102 | ||
| AK082785.1 | - | 3 - 4 | 286 | 102 | ||
| AK129211.1 | - | 12 - 13 | 1492 | 102 | ||
| AK132697.1 | - | 13 - 14 | 1543 | 102 | ||
| BC058809.1 | - | 12 - 13 | 2903 | 102 | ||
| BC099529.1 | - | 13 - 14 | 1566 | 102 | ||
| BC132195.1 | - | 13 - 14 | 1531 | 40130472 | 102 | |
| BC138153.1 | - | 13 - 14 | 1531 | 8861172 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ufl1
Genomic Map