Assay Details
| Gene Symbol: Ufl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026194.4 | NP_080470.2 | 2 - 3 | 290 | 154 | |
| GenBank mRNA | AK007993.2 | - | 2 - 3 | 235 | 154 | |
| AK032663.1 | - | 2 - 3 | 226 | 154 | ||
| AK076206.1 | - | 2 - 3 | 235 | 154 | ||
| AK132697.1 | - | 2 - 3 | 254 | 154 | ||
| BC058809.1 | - | 2 - 3 | 229 | 154 | ||
| BC080203.1 | - | 2 - 3 | 268 | 154 | ||
| BC132195.1 | - | 2 - 3 | 242 | 40130472 | 154 | |
| BC138153.1 | - | 2 - 3 | 242 | 8861172 | 154 | |
| BY097965.1 | - | 2 - 3 | 292 | 154 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ufl1
Genomic Map