Assay Details
| Gene Symbol: Tmprss11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030297.2 | NP_001025468.1 | 7 - 8 | 779 | 128 | |
| GenBank mRNA | AM260217.1 | - | 7 - 8 | 779 | 128 | |
| BC147157.1 | - | 7 - 8 | 760 | 9056205 | 128 | |
| BC147158.1 | - | 7 - 8 | 760 | 9056165 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss11c
Genomic Map