Assay Details
| Gene Symbol: Anxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010730.2 | NP_034860.2 | 6 - 7 | 541 | 118 | |
| GenBank mRNA | AK002681.1 | - | 6 - 7 | 543 | 118 | |
| AK140838.1 | - | 6 - 7 | 544 | 118 | ||
| AK150564.1 | - | 6 - 7 | 543 | 118 | ||
| AK150718.1 | - | 6 - 7 | 543 | 118 | ||
| AK153156.1 | - | 6 - 7 | 543 | 118 | ||
| AK153490.1 | - | 6 - 7 | 543 | 118 | ||
| AK167612.1 | - | 6 - 7 | 543 | 118 | ||
| CT009771.1 | - | 5 - 6 | 467 | 118 | ||
| EU684130.1 | - | 5 - 6 | 467 | 118 | ||
| M24554.1 | - | 5 - 6 | 452 | 118 | ||
| X07486.1 | - | 5 - 6 | 480 | 118 | ||
More Information
Set Membership: |
|

Anxa1
Genomic Map