Assay Details
| Gene Symbol: Cct3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009836.1 | NP_033966.1 | 1 - 2 | 125 | 71 | |
| GenBank mRNA | AK089548.1 | - | 1 - 2 | 135 | 71 | |
| AK146360.1 | - | 1 - 2 | 135 | 71 | ||
| AK154043.1 | - | 1 - 2 | 108 | 71 | ||
| AK156836.1 | - | 1 - 2 | 74 | 71 | ||
| AK158254.1 | - | 1 - 2 | 108 | 71 | ||
| AK158272.1 | - | 1 - 2 | 74 | 71 | ||
| AK161630.1 | - | 1 - 2 | 135 | 71 | ||
| AK171160.1 | - | 1 - 2 | 135 | 71 | ||
| BC054776.1 | - | 1 - 2 | 124 | 71 | ||
| BC139465.1 | - | 1 - 2 | 114 | 8862487 | 71 | |
| BC139467.1 | - | 1 - 2 | 114 | 8862489 | 71 | |
| Z31556.1 | - | 1 - 2 | 125 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK146360.1,AK154043.1,AK156836.1,AK171160.1,BC139465.1,BC139467.1,NM_009836.1,Z31556.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cct3
Genomic Map