Assay Details
| Gene Symbol: Paqr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198410.3 | NP_940802.1 | 4 - 5 | 444 | 81 | |
| GenBank mRNA | AK139144.1 | - | 4 - 5 | 445 | 81 | |
| AY424295.1 | - | 4 - 5 | 445 | 81 | ||
| BC120512.1 | - | 3 - 4 | 407 | 40129435 | 81 | |
| BC125428.1 | - | 3 - 4 | 407 | 40129943 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Paqr6
Genomic Map