Assay Details
| Gene Symbol: Trpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011643.2 | NP_035773.1 | 2 - 3 | 843 | 59 | |
| GenBank mRNA | BC137581.1 | - | 2 - 3 | 829 | 8860600 | 59 |
| U73625.1 | - | 2 - 3 | 843 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trpc1
Genomic Map