Assay Details
| Gene Symbol: Zfp451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290699.1 | NP_001277628.1 | 2 - 3 | 342 | 98 | |
| NM_001290700.1 | NP_001277629.1 | 2 - 3 | 342 | 98 | ||
| NM_133817.3 | NP_598578.1 | 2 - 3 | 342 | 98 | ||
| GenBank mRNA | AK015797.1 | - | 2 - 3 | 317 | 98 | |
| AK030088.1 | - | 2 - 3 | 175 | 98 | ||
| AK122541.1 | - | 2 - 3 | 167 | 98 | ||
| AK210231.1 | - | 1 - 2 | 89 | 98 | ||
| BC019546.1 | - | 2 - 3 | 178 | 4038791 | 98 | |
| BC062154.1 | - | 2 - 3 | 132 | 6331597 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC062154.1, NM_001290699.1 transcripts.
Set Membership: |
|

Zfp451
Genomic Map