Assay Details
| Gene Symbol: A630033H20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122596.1 | NP_001116068.1 | 1 - 2 | 221 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001122596.1 transcripts.
Set Membership: |
|

A630033H20Rik
Genomic Map