Assay Details
| Gene Symbol: Pik3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029094.3 | NP_083370.2 | 16 - 17 | 2472 | 113 | |
| GenBank mRNA | AK081350.1 | - | 16 - 17 | 2663 | 113 | |
| AK090116.1 | - | 16 - 17 | 2349 | 113 | ||
| AK154106.1 | - | 16 - 17 | 2457 | 113 | ||
| BC127073.1 | - | 15 - 16 | 2121 | 40110218 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pik3cb
Genomic Map