Assay Details
| Gene Symbol: Pik3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029094.3 | NP_083370.2 | 17 - 18 | 2635 | 81 | |
| GenBank mRNA | AK003230.1 | - | 1 - 2 | 50 | 81 | |
| AK081350.1 | - | 17 - 18 | 2826 | 81 | ||
| AK090116.1 | - | 17 - 18 | 2512 | 81 | ||
| AK154106.1 | - | 17 - 18 | 2620 | 81 | ||
| AK165230.1 | - | 95 | 81 | |||
| BC127073.1 | - | 16 - 17 | 2284 | 40110218 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pik3cb
Genomic Map