Assay Details
| Gene Symbol: Pibf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029320.3 | NP_083596.2 | 11 - 12 | 2210 | 104 | |
| GenBank mRNA | AK006060.1 | - | 2 - 3 | 184 | 104 | |
| AK045133.1 | - | 11 - 12 | 1707 | 104 | ||
| AK149007.1 | - | 6 - 7 | 763 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006060.1 transcripts.
Set Membership: |
|

Pibf1
Genomic Map