Assay Details
| Gene Symbol: Pibf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029320.3 | NP_083596.2 | 13 - 14 | 2461 | 134 | |
| GenBank mRNA | AK006060.1 | - | 4 - 5 | 435 | 134 | |
| AK017117.1 | - | 4 - 5 | 527 | 134 | ||
| AK045133.1 | - | 13 - 14 | 1958 | 134 | ||
| AK149007.1 | - | 8 - 9 | 1014 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pibf1
Genomic Map