Assay Details
| Gene Symbol: Pibf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029320.3 | NP_083596.2 | 5 - 6 | 1393 | 69 | |
| GenBank mRNA | AK030385.1 | - | 5 - 6 | 874 | 69 | |
| AK045133.1 | - | 5 - 6 | 888 | 69 | ||
| BC113804.1 | - | 5 - 6 | 870 | 69 | ||
| BC129811.1 | - | 5 - 6 | 870 | 40062382 | 69 | |
| BC129812.1 | - | 5 - 6 | 870 | 40062383 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pibf1
Genomic Map