Assay Details
| Gene Symbol: Pibf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029320.3 | NP_083596.2 | 6 - 7 | 1536 | 80 | |
| GenBank mRNA | AK030385.1 | - | 6 - 7 | 1017 | 80 | |
| AK045133.1 | - | 6 - 7 | 1031 | 80 | ||
| AK149007.1 | - | 1 - 2 | 89 | 80 | ||
| BC113804.1 | - | 6 - 7 | 1013 | 80 | ||
| BC129811.1 | - | 6 - 7 | 1013 | 40062382 | 80 | |
| BC129812.1 | - | 6 - 7 | 1013 | 40062383 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK149007.1 transcripts.
Set Membership: |
|

Pibf1
Genomic Map