Assay Details
| Gene Symbol: Pibf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029320.3 | NP_083596.2 | 7 - 8 | 1648 | 82 | |
| GenBank mRNA | AK030385.1 | - | 7 - 8 | 1129 | 82 | |
| AK045133.1 | - | 7 - 8 | 82 | |||
| AK149007.1 | - | 2 - 3 | 201 | 82 | ||
| BC113804.1 | - | 7 - 8 | 1125 | 82 | ||
| BC129811.1 | - | 7 - 8 | 1125 | 40062382 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK149007.1 transcripts.
Set Membership: |
|

Pibf1
Genomic Map