Assay Details
| Gene Symbol: Pibf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029320.3 | NP_083596.2 | 9 - 10 | 1945 | 68 | |
| GenBank mRNA | AK030385.1 | - | 9 - 10 | 1426 | 68 | |
| AK045133.1 | - | 9 - 10 | 1442 | 68 | ||
| AK149007.1 | - | 4 - 5 | 498 | 68 | ||
| BC129811.1 | - | 9 - 10 | 1422 | 40062382 | 68 | |
| BC129812.1 | - | 9 - 10 | 1422 | 40062383 | 68 | |
More Information
Set Membership: |
|

Pibf1
Genomic Map