Assay Details
| Gene Symbol: Atp6v1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133826.4 | NP_598587.2 | 8 - 9 | 818 | 139 | |
| GenBank mRNA | AK034165.1 | - | 6 - 7 | 2977 | 139 | |
| AK078767.1 | - | 8 - 9 | 819 | 139 | ||
| AK152147.1 | - | 8 - 9 | 840 | 139 | ||
| BC009154.1 | - | 8 - 9 | 819 | 3601621 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1h
Genomic Map