Assay Details
| Gene Symbol: Cops5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277101.1 | NP_001264030.1 | 5 - 6 | 753 | 155 | |
| NM_013715.2 | NP_038743.1 | 5 - 6 | 1002 | 155 | ||
| NR_102282.1 | 5 - 6 | 1010 | 155 | |||
| GenBank mRNA | AF065386.1 | - | 5 - 6 | 755 | 155 | |
| AF068223.1 | - | 5 - 6 | 770 | 155 | ||
| AK002761.1 | - | 5 - 6 | 783 | 155 | ||
| AK012499.1 | - | 5 - 6 | 789 | 155 | ||
| AK146271.1 | - | 5 - 6 | 789 | 155 | ||
| AK151492.1 | - | 5 - 6 | 788 | 155 | ||
| BC046753.1 | - | 5 - 6 | 748 | 5693100 | 155 | |
| DV056728.1 | - | 5 - 6 | 809 | 155 | ||
| U70736.1 | - | 5 - 6 | 778 | 155 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cops5
Genomic Map