Assay Details
| Gene Symbol: Cops5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277101.1 | NP_001264030.1 | 6 - 7 | 861 | 153 | |
| NM_013715.2 | NP_038743.1 | 6 - 7 | 1110 | 153 | ||
| NR_102282.1 | 6 - 7 | 1118 | 153 | |||
| GenBank mRNA | AF065386.1 | - | 6 - 7 | 863 | 153 | |
| AF068223.1 | - | 6 - 7 | 878 | 153 | ||
| AK002761.1 | - | 6 - 7 | 891 | 153 | ||
| AK012499.1 | - | 6 - 7 | 897 | 153 | ||
| AK146271.1 | - | 6 - 7 | 897 | 153 | ||
| AK151492.1 | - | 6 - 7 | 896 | 153 | ||
| BC046753.1 | - | 6 - 7 | 856 | 5693100 | 153 | |
| U70736.1 | - | 6 - 7 | 886 | 153 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cops5
Genomic Map