Assay Details
| Gene Symbol: Ubac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026861.2 | NP_081137.2 | 3 - 4 | 419 | 97 | |
| GenBank mRNA | AK049109.1 | - | 3 - 4 | 368 | 97 | |
| AK158177.1 | - | 3 - 4 | 368 | 97 | ||
| AK199665.1 | - | 1 - 2 | 105 | 97 | ||
| BC024467.1 | - | 3 - 4 | 405 | 4975777 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubac2
Genomic Map