Assay Details
| Gene Symbol: Itgb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177290.3 | NP_796264.2 | ||||
| GenBank mRNA | AK029281.1 | - | ||||
| AK053056.1 | - | |||||
| BC120691.1 | - | 40129614 | ||||
| BC125343.1 | - | 40129858 | ||||
| BC144785.1 | - | |||||

Itgb8
Genomic Map