Assay Details
| Gene Symbol: Wdr60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146039.3 | NP_666151.3 | 9 - 10 | 1582 | 149 | |
| GenBank mRNA | AK052482.1 | - | 9 - 10 | 1546 | 149 | |
| AK144138.1 | - | 9 - 10 | 1582 | 149 | ||
More Information
Set Membership: |
|

Wdr60
Genomic Map