Assay Details
| Gene Symbol: Wdr60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146039.3 | NP_666151.3 | 10 - 11 | 1692 | 79 | |
| GenBank mRNA | AK052482.1 | - | 10 - 11 | 1656 | 79 | |
| AK144138.1 | - | 10 - 11 | 1692 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wdr60
Genomic Map