Assay Details
| Gene Symbol: Gabra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010250.5 | NP_034380.1 | 1 - 2 | 767 | 99 | |
| GenBank mRNA | AK034095.1 | - | 1 - 2 | 768 | 99 | |
| AK141596.1 | - | 1 - 2 | 627 | 99 | ||
| BC132331.1 | - | 1 - 2 | 443 | 40130608 | 99 | |
| BC137733.1 | - | 1 - 2 | 446 | 8860752 | 99 | |
| M63436.1 | - | 1 - 2 | 440 | 99 | ||
| M86566.1 | - | 1 - 2 | 135 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK034095.1, AK141596.1, BC132331.1, BC137733.1, M63436.1, M86566.1, NM_010250.5 transcripts.
Set Membership: |
|

Gabra1
Genomic Map