Assay Details
| Gene Symbol: Chpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146690.1 | NP_001140162.1 | 3 - 4 | 675 | 77 | |
| NM_144807.3 | NP_659056.3 | 3 - 4 | 675 | 77 | ||
| NR_027477.1 | 3 - 4 | 675 | 77 | |||
| GenBank mRNA | AK007964.1 | - | 3 - 4 | 595 | 77 | |
| AK032497.1 | - | 3 - 4 | 570 | 77 | ||
| AY445814.1 | - | 3 - 4 | 570 | 77 | ||
| EL958096.1 | - | 2 - 3 | 275 | 77 | ||
More Information
Set Membership: |
|

Chpt1
Genomic Map