Assay Details
| Gene Symbol: Ccdc53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027487.1 | NP_081763.1 | 5 - 6 | 585 | 76 | |
| GenBank mRNA | AK017732.1 | - | 5 - 6 | 513 | 76 | |
More Information
Set Membership: |
|

Ccdc53
Genomic Map