Assay Details
| Gene Symbol: Armc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081393.1 | NP_001074862.1 | 14 - 15 | 2105 | 135 | |
| GenBank mRNA | BC158097.1 | - | 13 - 14 | 2094 | 9088178 | 135 |
More Information
Updated Mapping Information
Set Membership: |
|

Armc4
Genomic Map