Assay Details
| Gene Symbol: Qpct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027455.2 | NP_081731.1 | 4 - 5 | 1086 | 83 | |
| GenBank mRNA | AK017598.1 | - | 4 - 5 | 749 | 83 | |
| AK037197.1 | - | 4 - 5 | 739 | 83 | ||
| AK045974.1 | - | 4 - 5 | 1087 | 83 | ||
| AK148394.1 | - | 4 - 5 | 757 | 83 | ||
| BC020023.1 | - | 3 - 4 | 657 | 3491756 | 83 | |
| BC151027.1 | - | 4 - 5 | 830 | 9087940 | 83 | |
| BC151029.1 | - | 4 - 5 | 830 | 9087942 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Qpct
Genomic Map