Assay Details
| Gene Symbol: Abcb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029961.2 | NP_084237.1 | 13 - 14 | 1617 | 84 | |
| GenBank mRNA | AY766239.1 | - | 13 - 14 | 1581 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcb5
Genomic Map