Assay Details
| Gene Symbol: Abcb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029961.2 | NP_084237.1 | 21 - 22 | 2716 | 99 | |
| GenBank mRNA | AK139832.1 | - | 8 - 9 | 1110 | 99 | |
| AY766238.1 | - | 1 - 2 | 213 | 99 | ||
| AY766239.1 | - | 21 - 22 | 2680 | 99 | ||
| JQ655148.1 | - | 8 - 9 | 2195 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb5
Genomic Map