Assay Details
| Gene Symbol: Abcb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029961.2 | NP_084237.1 | 1 - 2 | 66 | 114 | |
| GenBank mRNA | AK020318.1 | - | 1 - 2 | 67 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK020318.1, NM_029961.2 transcripts.
Set Membership: |
|

Abcb5
Genomic Map