Assay Details
| Gene Symbol: Abcb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029961.2 | NP_084237.1 | 24 - 25 | 3109 | 85 | |
| GenBank mRNA | AY766239.1 | - | 24 - 25 | 3073 | 85 | |
| JQ655148.1 | - | 11 - 12 | 2588 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb5
Genomic Map