Assay Details
| Gene Symbol: Abcb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029961.2 | NP_084237.1 | 3 - 4 | 196 | 74 | |
| GenBank mRNA | AK020318.1 | - | 3 - 4 | 197 | 74 | |
| AY766239.1 | - | 3 - 4 | 160 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb5
Genomic Map