Assay Details
| Gene Symbol: Slc7a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK077593.1 | - | 10 - 11 | 1759 | 56 | |
| AK141558.1 | - | 9 - 10 | 2231 | 56 | ||
| AK165931.1 | - | 10 - 11 | 1756 | 56 | ||
| BC050195.1 | - | 10 - 11 | 1735 | 30010078 | 56 | |
| U70859.1 | - | 11 - 12 | 1893 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a3
Genomic Map