Assay Details
| Gene Symbol: L3mbtl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177278.5 | NP_796252.2 | 12 - 13 | 1563 | 95 | |
| GenBank mRNA | AK053068.1 | - | 11 - 12 | 1478 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

L3mbtl4
Genomic Map