Assay Details
| Gene Symbol: Dnajc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008929.3 | NP_032955.2 | 10 - 11 | 1360 | 135 | |
| GenBank mRNA | AK004033.1 | - | 5 - 6 | 661 | 135 | |
| AK146097.1 | - | 10 - 11 | 1324 | 135 | ||
| AK148270.1 | - | 10 - 11 | 1317 | 135 | ||
| BC013766.1 | - | 10 - 11 | 1305 | 2646147 | 135 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc3
Genomic Map