Assay Details
| Gene Symbol: Dnajc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008929.3 | NP_032955.2 | 5 - 6 | 701 | 70 | |
| GenBank mRNA | AK146097.1 | - | 5 - 6 | 665 | 70 | |
| AK148270.1 | - | 5 - 6 | 658 | 70 | ||
| U28423.1 | - | 5 - 6 | 644 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc3
Genomic Map