Assay Details
| Gene Symbol: Ppp1r12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027892.2 | NP_082168.1 | 9 - 10 | 1244 | 90 | |
| GenBank mRNA | AK004785.1 | - | 9 - 10 | 1244 | 90 | |
| AK139570.1 | - | 9 - 10 | 1130 | 90 | ||
| BC125381.1 | - | 10 - 11 | 1313 | 40129896 | 90 | |
| BC137630.1 | - | 10 - 11 | 1313 | 8860649 | 90 | |
More Information
Set Membership: |
|

Ppp1r12a
Genomic Map