Assay Details
| Gene Symbol: Eml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114361.1 | NP_001107833.1 | 19 - 20 | 2375 | 125 | |
| NM_001114362.1 | NP_001107834.1 | 17 - 18 | 2168 | 125 | ||
| NM_001286567.1 | NP_001273496.1 | 19 - 20 | 2278 | 125 | ||
| NM_199466.3 | NP_955760.3 | 16 - 17 | 2039 | 125 | ||
| GenBank mRNA | AK077527.1 | - | 11 - 12 | 1204 | 125 | |
| AK139067.1 | - | 3 - 4 | 232 | 125 | ||
| AK144604.1 | - | 19 - 20 | 2376 | 125 | ||
| AK189201.1 | - | 2 - 3 | 127 | 125 | ||
| BC032258.1 | - | 5 - 6 | 523 | 125 | ||
| BC065178.1 | - | 19 - 20 | 2216 | 6827397 | 125 | |
| BC066099.1 | - | 16 - 17 | 1970 | 30533573 | 125 | |
| BC067011.1 | - | 16 - 17 | 1988 | 6405184 | 125 | |
| BC070427.1 | - | 17 - 18 | 2108 | 6413041 | 125 | |
| BC079619.1 | - | 16 - 17 | 1970 | 30533573 | 125 | |
| DQ230326.1 | - | 18 - 19 | 2058 | 125 | ||
More Information
Set Membership: |
|

Eml4
Genomic Map