Assay Details
| Gene Symbol: Abcg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031884.1 | NP_114090.1 | 7 - 8 | 956 | 72 | |
| GenBank mRNA | AF312713.2 | - | 7 - 8 | 1044 | 72 | |
| AK149569.1 | - | 7 - 8 | 927 | 72 | ||
| BC106766.2 | - | 7 - 8 | 933 | 40039484 | 72 | |
More Information
Set Membership: |
|

Abcg5
Genomic Map