Assay Details
| Gene Symbol: Arhgef4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183019.2 | NP_898840.2 | 4 - 5 | 821 | 83 | |
| GenBank mRNA | AK034013.1 | - | 4 - 5 | 785 | 83 | |
| AK134629.1 | - | 784 | 83 | |||
| AK138598.1 | - | 4 - 5 | 781 | 83 | ||
| AK163424.1 | - | 4 - 5 | 803 | 83 | ||
| AK169449.1 | - | 4 - 5 | 804 | 83 | ||
| AK169957.1 | - | 4 - 5 | 841 | 83 | ||
| BC055804.1 | - | 4 - 5 | 783 | 83 | ||
| BC094608.1 | - | 4341 | 83 | |||
| BC139371.1 | - | 4 - 5 | 804 | 8862393 | 83 | |
| BC139397.1 | - | 4 - 5 | 804 | 8862419 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef4
Genomic Map