Assay Details
| Gene Symbol: Sgk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037759.1 | NP_001032848.1 | 10 - 11 | 962 | 115 | |
| NM_133220.2 | NP_573483.1 | 11 - 12 | 1081 | 115 | ||
| NM_177547.3 | NP_808215.2 | 10 - 11 | 974 | 115 | ||
| GenBank mRNA | AF312007.1 | - | 11 - 12 | 1023 | 115 | |
| AK028722.1 | - | 11 - 12 | 1026 | 115 | ||
| AK030314.1 | - | 10 - 11 | 963 | 115 | ||
| AK031133.1 | - | 11 - 12 | 1062 | 115 | ||
| AK031328.1 | - | 10 - 11 | 903 | 115 | ||
| AK146561.1 | - | 10 - 11 | 917 | 115 | ||
| AK151181.1 | - | 11 - 12 | 1026 | 115 | ||
| AK151793.1 | - | 10 - 11 | 954 | 115 | ||
| AK151867.1 | - | 11 - 12 | 1058 | 115 | ||
| AK151996.1 | - | 11 - 12 | 1225 | 115 | ||
| AK166414.1 | - | 10 - 11 | 904 | 115 | ||
| AK166531.1 | - | 10 - 11 | 923 | 115 | ||
| AK171120.1 | - | 10 - 11 | 950 | 115 | ||
| AK171186.1 | - | 10 - 11 | 950 | 115 | ||
| AK171678.1 | - | 10 - 11 | 934 | 115 | ||
| AK171959.1 | - | 10 - 11 | 1011 | 115 | ||
| AK172323.1 | - | 10 - 11 | 924 | 115 | ||
| BC018363.1 | - | 8 - 9 | 612 | 2631682 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgk3
Genomic Map