Assay Details
| Gene Symbol: Sgk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037759.1 | NP_001032848.1 | 11 - 12 | 1078 | 102 | |
| NM_133220.2 | NP_573483.1 | 12 - 13 | 1197 | 102 | ||
| NM_177547.3 | NP_808215.2 | 11 - 12 | 1090 | 102 | ||
| GenBank mRNA | AF312007.1 | - | 12 - 13 | 1139 | 102 | |
| AK028722.1 | - | 12 - 13 | 1142 | 102 | ||
| AK030314.1 | - | 11 - 12 | 1079 | 102 | ||
| AK031133.1 | - | 12 - 13 | 1178 | 102 | ||
| AK031328.1 | - | 11 - 12 | 1019 | 102 | ||
| AK083319.1 | - | 2 - 3 | 145 | 102 | ||
| AK143836.1 | - | 12 - 13 | 1185 | 102 | ||
| AK146561.1 | - | 11 - 12 | 1033 | 102 | ||
| AK151181.1 | - | 12 - 13 | 1142 | 102 | ||
| AK151793.1 | - | 11 - 12 | 1070 | 102 | ||
| AK151867.1 | - | 12 - 13 | 1174 | 102 | ||
| AK151996.1 | - | 12 - 13 | 1341 | 102 | ||
| AK166414.1 | - | 11 - 12 | 1020 | 102 | ||
| AK166531.1 | - | 11 - 12 | 1039 | 102 | ||
| AK171120.1 | - | 11 - 12 | 1066 | 102 | ||
| AK171186.1 | - | 11 - 12 | 1066 | 102 | ||
| AK171678.1 | - | 11 - 12 | 1050 | 102 | ||
| AK171959.1 | - | 11 - 12 | 1127 | 102 | ||
| AK172323.1 | - | 11 - 12 | 1040 | 102 | ||
| AK196812.1 | - | 1 - 2 | 50 | 102 | ||
| BC018363.1 | - | 9 - 10 | 728 | 2631682 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgk3
Genomic Map