Assay Details
| Gene Symbol: Sgk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037759.1 | NP_001032848.1 | 14 - 15 | 1300 | 62 | |
| NM_133220.2 | NP_573483.1 | 15 - 16 | 1419 | 62 | ||
| NM_177547.3 | NP_808215.2 | 14 - 15 | 1312 | 62 | ||
| GenBank mRNA | AF312007.1 | - | 15 - 16 | 1361 | 62 | |
| AK028722.1 | - | 14 - 15 | 60 | |||
| AK030314.1 | - | 14 - 15 | 1301 | 62 | ||
| AK031133.1 | - | 15 - 16 | 1400 | 62 | ||
| AK031328.1 | - | 14 - 15 | 1241 | 62 | ||
| AK143836.1 | - | 15 - 16 | 1465 | 62 | ||
| AK146561.1 | - | 14 - 15 | 1255 | 62 | ||
| AK151181.1 | - | 15 - 16 | 1364 | 62 | ||
| AK151867.1 | - | 15 - 16 | 1396 | 62 | ||
| AK151996.1 | - | 15 - 16 | 1563 | 62 | ||
| AK166531.1 | - | 14 - 15 | 1261 | 62 | ||
| AK171120.1 | - | 14 - 15 | 1288 | 62 | ||
| AK171186.1 | - | 14 - 15 | 1288 | 62 | ||
| AK171678.1 | - | 14 - 15 | 1272 | 62 | ||
| AK171959.1 | - | 14 - 15 | 1349 | 62 | ||
| AK172323.1 | - | 14 - 15 | 1262 | 62 | ||
| BC018363.1 | - | 12 - 13 | 950 | 2631682 | 62 | |
More Information
Set Membership: |
|

Sgk3
Genomic Map